Check Order Status
Orthophosphoric Acid, 85+%, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Orthophosphoric Acid, 85+%, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
500 mL
2.5 L
Packaging:
HDPE plastic bottle
Catalog number
FKCO0450PB15
also known as FKCO/0450/PB15
Price (MYR)
159.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
159.00
EA
Add to cart
Specifications
Boiling Point
158°C
Melting Point
21°C
CAS
7664-38-2
pH
2
Physical Form
Liquid
View more