Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
HDPE plastic bottle
Catalog number FKCN2300PB15
also known as FKCN/2300/PB15
Price (MYR)
150.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
150.00
EA
Add to cart
Specifications
pH1
ColorColorless Yellow
CAS7697-37-2
Melting Point-42°C
Boiling Point122°C
View more
HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012