Check Order Status
Molecular Sieve Type 4A, Technical, 1.6-2.5 mm Beads (8-12 Mesh), Fisher Chemical™
Element Search
Structure Search
Molecular Sieve Type 4A, Technical, 1.6-2.5 mm Beads (8-12 Mesh), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
Catalog number
FKCMS103053
also known as FKCMS/1030/53
Price (MYR)
205.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
205.00
EA
Add to cart
Specifications
Color
Beige
Packaging
Plastic powder jar
Quantity
500 g
pH
11
CAS
70955-01-0
View more