Check Order Status
Molecular Sieve Type 4A, Technical, 2.5-5.0 mm Beads (4-8 Mesh), Fisher Chemical™
Element Search
Structure Search
Molecular Sieve Type 4A, Technical, 2.5-5.0 mm Beads (4-8 Mesh), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
Catalog number
FKCMS102053
also known as FKCMS/1020/53
Price (MYR)
213.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
213.00
BOT
Add to cart
Specifications
Color
Beige
Packaging
Plastic powder jar
Quantity
500 g
pH
11
CAS
70955-01-0
View more