Check Order Status
Molecular Sieve Type 3A, Technical, 1.6-2.5mm Beads (8-12 Mesh), Fisher Chemical™
Actual product may vary
Molecular Sieve Type 3A, Technical, 1.6-2.5mm Beads (8-12 Mesh), Fisher Chemical™
AlNaO6Si2, CAS Number-308080-99-1, 500 g, Beige, URGAHOPLAPQHLN-UHFFFAOYSA-N, aluminum;sodium;dioxido(oxo)silane, 131613, 202.135, Plastic powder jar, Solid, 19758701, [O-][Si](-O)[O-].[O-][Si](-O)[O-].[Na+].[Al+3] | CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Catalog Number
Quantity
FKCMS101053
also known as FKCMS/1010/53
500 g
Catalog number
FKCMS101053
also known as FKCMS/1010/53
Price (MYR)
242.00
EA
Add to cart
Quantity:
500 g
Price (MYR)
242.00
EA
Add to cart
AlNaO6Si2, CAS Number-308080-99-1, 500 g, Beige, URGAHOPLAPQHLN-UHFFFAOYSA-N, aluminum;sodium;dioxido(oxo)silane, 131613, 202.135, Plastic powder jar, Solid, 19758701, [O-][Si](-O)[O-].[O-][Si](-O)[O-].[Na+].[Al+3]
Specifications
Color
Beige
Packaging
Plastic powder jar
Quantity
500 g
CAS
308080-99-1
Physical Form
Solid
Molecular Formula
AlNaO6Si2
MDL Number
MFCD00131613
InChI Key
URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
IUPAC Name
aluminum;sodium;dioxido(oxo)silane
Molecular Weight (g/mol)
202.135
PubChem CID
19758701