Check Order Status
1-Methyl-2-Pyrrolidinone, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
1-Methyl-2-Pyrrolidinone, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
500 mL
Packaging:
View all
Metal Can
Catalog number
FKCM512525
also known as FKCM/5125/25
Price (MYR)
-
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Specifications
pH
7.7 to 8.0
Melting Point
-24°C
Color
Colorless
Boiling Point
202°C
Quantity
25 L
View more