Methanol, Extra Dry, for Synthesis, Fisher Chemical™
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
Amber Glass Bottle
Catalog number FKCM405017
also known as FKCM/4050/17
Price (MYR)
130.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
130.00
BOT
Add to cart
Specifications
Melting Point-98°C
CAS67-56-1
ColorColorless
Boiling Point64.7°C
Quantity2.5 L
View more
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042