Check Order Status
Methanol, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Methanol, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
Packaging:
Metal Can
Catalog number
FKCM395025
also known as FKCM/3950/25
Price (MYR)
762.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
762.00
EA
Add to cart
Specifications
Melting Point
-98°C
CAS
67-56-1
Color
Colorless
Boiling Point
64.7°C
Quantity
25 L
View more