Check Order Status
Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™
Actual product may vary
Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™
HO3P, CAS Number-37267-86-0, unii-mtk99r3uv0, x-phosphate-group, m-phosphoric acid, metaphosphoric acid hpo3, meta-phosphoric acid, phosphenic acid, metaphosphoric acid, phosphonate 1-, hydrogenphosphonate, hydrogen phosphonate, 500g, Colorless, UEZVMMHDMIWARA-UHFFFAOYSA-N, 200 deg.C, 11351, 79.979 | CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FKCM385153
also known as FKCM/3851/53
500 g
Plastic powder jar
Catalog number
FKCM385153
also known as FKCM/3851/53
Price (MYR)
1,041.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Price (MYR)
1,041.00
BOT
Add to cart
HO3P, CAS Number-37267-86-0, unii-mtk99r3uv0, x-phosphate-group, m-phosphoric acid, metaphosphoric acid hpo3, meta-phosphoric acid, phosphenic acid, metaphosphoric acid, phosphonate 1-, hydrogenphosphonate, hydrogen phosphonate, 500g, Colorless, UEZVMMHDMIWARA-UHFFFAOYSA-N, 200 deg.C, 11351, 79.979
Specifications
pH
2
Color
Colorless
CAS
37267-86-0
Melting Point
200°C
Packaging
Plastic powder jar
Quantity
500 g
Molecular Formula
HO3P
Physical Form
Solid
MDL Number
11351
Synonym
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
InChI Key
UEZVMMHDMIWARA-UHFFFAOYSA-N
SMILES
OP(=O)=O
Molecular Weight (g/mol)
79.979
PubChem CID
3084658