Check Order Status
Lead(II) Acetate Trihydrate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Lead(II) Acetate Trihydrate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 kg
Catalog number
FKCL095060
also known as FKCL/0950/60
Price (MYR)
1,153.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,153.00
EA
Add to cart
Specifications
CAS
6080-56-4
Melting Point
75°C
Color
White
pH
5.5 to 6.5
Packaging
Plastic powder jar
View more