Check Order Status
Lactic Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Element Search
Structure Search
Lactic Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FKCL0150PB17
also known as FKCL/0150/PB17
Price (MYR)
884.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
884.00
EA
Add to cart
Specifications
pH
1,2
CAS
79-33-4
Boiling Point
125°C
Color
Yellow
Molecular Formula
C3H6O3
View more
Lactic Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur