Check Order Status
Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical
Anolyte solution for general use in conventional cells with a diaphragm
Have Questions?
Catalog Number
Quantity
FKCK250008
also known as FKCK/2500/08
500 mL
Catalog number
FKCK250008
also known as FKCK/2500/08
Price (MYR)
355.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
355.00
EA
Add to cart
Chemical Identifiers
CAS
67-66-3, 108-75-8, 111-42-2, 7446-09-5
Specifications
Chemical Name or Material
Aqualine™ Electrolyte A
Color
Colorless
Quantity
500 mL
Name Note
Karl Fischer Reagent
ChemAlert Storage Symbol
Red
View more
Anolyte solution for use in conventional cells with a diaphragm
Source of electrolyte required for anodic oxidation for coulometric analysis
Contains methanol and chloroform as solvents