Check Order Status
Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Element Search
Structure Search
Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
500 mL
Catalog number
FKCK2250R15
also known as FKCK/2250R/15
Price (MYR)
557.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
557.00
EA
Add to cart
Chemical Identifiers
CAS
111-90-0, 5683-33-0, 7446-09-5, 7553-56-2
Specifications
Chemical Name or Material
Aqualine™ Complete 5K
Color
Colorless
Quantity
1 L
Cleanup Symbol
Solvent
Name Note
Karl Fischer Reagent
View more
Single-component volumetric reagent for high water content analysis of aldehydes and ketones
Water equivalent: 5.0mg H
2
O/mL
Unlimited water capacity
Perform titer determination before use
Pyridine free