Check Order Status
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
For two-component system. Water equivalent: 5.0mg H
2
O/mL
Have Questions?
Catalog Number
Quantity
FKCK220015
also known as FKCK/2200/15
1 L
Catalog number
FKCK220015
also known as FKCK/2200/15
Price (MYR)
328.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
328.00
EA
Add to cart
Specifications
Chemical Name or Material
Aqualine™ Titrant 5
Color
Colorless
Purity or Free of Statement
Pyridine free
Quantity
1 L
Cleanup Symbol
Solvent
View more