Check Order Status
Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Element Search
Structure Search
Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Single-component reagent; Pyridine free
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCK200017
also known as FKCK/2000/17
Price (MYR)
990.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
990.00
BOT
Add to cart
Chemical Identifiers
CAS
111-90-0, 288-32-4, 7446-09-5, 7553-09-5
Specifications
Chemical Name or Material
Aqualine™ Complete 5
Color
Brown
Purity or Free of Statement
Pyridine free
Quantity
2.5 L
Cleanup Symbol
Solvent
View more
Single-component reagent; Pyridine free
Single-component volumetric reagent for high water content analysis
Water equivalent: 5.0mg H
2
O/mL
Unlimited water capacity
Perform titer determination before use
Pyridine free