Check Order Status
Kjeldahl Boric Acid Solution, Pure, Solution 4% Alkaline with Indicator, Fisher Chemical™
Element Search
Structure Search
Kjeldahl Boric Acid Solution, Pure, Solution 4% Alkaline with Indicator, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
5 L
Catalog number
FKCK020015
also known as FKCK/0200/15
Price (MYR)
170.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
170.00
EA
Add to cart
Specifications
Color
Red
Quantity
1 L
Packaging
Plastic Container
Physical Form
Liquid
MDL Number
11337
View more