Check Order Status
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
Packaging:
HDPE plastic bottle
Catalog number
FKCJ846015
also known as FKCJ/8460/15
Price (MYR)
130.00
BOT
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
130.00
BOT
Add to cart
Specifications
Color
Colorless
CAS
7664-93-9
Boiling Point
100°C
Packaging
HDPE plastic bottle
Vapor Pressure
1mmHg at 20°C
View more