Check Order Status
Vanadium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Vanadium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Catalog number
FKCJ815008
also known as FKCJ/8150/08
Price (MYR)
539.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
539.00
EA
Add to cart
Specifications
Packaging
HDPE plastic bottle
Physical Form
Liquid
MDL Number
11453
Color
Colorless
Quantity
500 mL
View more