Check Order Status
Zinc Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Zinc Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FKCJ807008
also known as FKCJ/8070/08
500 mL
Catalog number
FKCJ807008
also known as FKCJ/8070/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Color
Colorless
pH
1
CAS
7779-88-6
Molecular Formula
N2O6Zn
MDL Number
11291
View more