Check Order Status
Sodium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Sodium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
100 mL
Catalog number
FKCJ806508
also known as FKCJ/8065/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Color
Colorless
pH
1
MDL Number
85307
Quantity
500 mL
Packaging
HDPE plastic bottle
View more
Sodium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M