Check Order Status
Lithium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Lithium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Catalog number
FKCJ803708
also known as FKCJ/8037/08
Price (MYR)
485.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
485.00
BOT
Add to cart
Specifications
Packaging
HDPE plastic bottle
Physical Form
Liquid
MDL Number
134051
Color
Colorless
pH
1
View more