Check Order Status
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
100 mL
Catalog number
FKCJ803008
also known as FKCJ/8030/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Color
Colorless
pH
1
CAS
10421-48-4
Molecular Formula
FeN3O9
MDL Number
11003
View more
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M