Check Order Status
Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Catalog number
FKCJ802508
also known as FKCJ/8025/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Color
Colorless
pH
1
CAS
3251-23-8
Molecular Formula
CuN2O6
MDL Number
10967
View more
Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M