Check Order Status
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
6 Ampules
1 Ampule
Catalog number
FKCJ7630C90
also known as FKCJ/7630C/90
Price (MYR)
837.00
EA
Add to cart
Quantity:
6 Ampules
Request bulk or custom format
Price (MYR)
837.00
EA
Add to cart
Specifications
Color
Colorless
MDL Number
3548
Quantity
6 Ampules
Packaging
Ampoule
Physical Form
Liquid
View more