Check Order Status
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
HDPE plastic bottle
Catalog number
FKCJ763015
also known as FKCJ/7630/15
Price (MYR)
380.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
380.00
EA
Add to cart
Specifications
pH
12
Color
Colorless
Boiling Point
100°C
CAS
1310-73-2
Melting Point
0°C
View more
Sodium Hydroxide Solution , NIST standard solution ready to use, for volumetric analysis