Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Actual product may vary
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
HNaO, CAS Number-1310-73-2, sodium hydroxide solution, ascarite, aetznatron, soda lye, sodium hydrate, caustic soda, sodium hydroxide na oh, sodium hydroxide, white caustic, soda, caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997 | CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantity
FKCJ762015
also known as FKCJ/7620/15
1 L
Catalog number FKCJ762015
also known as FKCJ/7620/15
Price (MYR)
306.00
BOT
Add to cart
Quantity:
1 L
Price (MYR)
306.00
BOT
Add to cart
HNaO, CAS Number-1310-73-2, sodium hydroxide solution, ascarite, aetznatron, soda lye, sodium hydrate, caustic soda, sodium hydroxide na oh, sodium hydroxide, white caustic, soda, caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
ColorColorless
Boiling Point100°C
CAS1310-73-2
Molecular FormulaHNaO
MDL Number3548
Synonymsodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity1 L
SMILES[OH-].[Na+]
IUPAC Namesodium;hydroxide
Molecular Weight (g/mol)39.997
PubChem CID14798
ChEBICHEBI:32145
PackagingHDPE plastic bottle
Vapor Pressure23 &20
Physical FormLiquid
Unit SizeBOT