Check Order Status
Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
HDPE plastic bottle
Catalog number
FKCJ7610PB15
also known as FKCJ/7610/PB15
Price (MYR)
206.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
206.00
EA
Add to cart
Specifications
pH
14
Color
Colorless
Boiling Point
105°C
CAS
1310-73-2
Melting Point
-10°C
View more
Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur.