Check Order Status
Potassium Hydroxide Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical
Element Search
Structure Search
Potassium Hydroxide Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FKCJ6630C90
also known as FKCJ/6630C/90
6 Ampules
Ampoule
Catalog number
FKCJ6630C90
also known as FKCJ/6630C/90
Price (MYR)
1,279.00
EA
Add to cart
Quantity:
6 Ampules
Packaging:
Ampoule
Request bulk or custom format
Price (MYR)
1,279.00
EA
Add to cart
Specifications
Color
Colorless
MDL Number
3553
Quantity
6 Ampules
Packaging
Ampoule
Physical Form
Liquid
View more