Check Order Status
Potassium Hydroxide Solution 0.1M (0.1N), (In Ethanol), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Potassium Hydroxide Solution 0.1M (0.1N), (In Ethanol), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
Catalog number
FKCJ662015
also known as FKCJ/6620/15
Price (MYR)
210.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
210.00
EA
Add to cart
Specifications
MDL Number
3553
Quantity
1 L
Packaging
HDPE plastic bottle
Physical Form
Liquid