Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
ClHO4, CAS Number-7601-90-3, unii-v561v90bg2, ueberchlorsaeure, perchlorsaeure, perchloric acid, hclo4, hyperchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchloric acid, 1L, CHEBI:29221, Colorless, VLTRZXGMWDSKGL-UHFFFAOYSA-N, 11325, 100.454
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FKCJ5820PB17
also known as FKCJ/5820/PB17
Price (MYR)
483.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
483.00
EA
Add to cart
Specifications
pH0.1
ColorColorless
CAS7601-90-3
PackagingHDPE plastic bottle
Quantity2.5 L
View more
ClHO4, CAS Number-7601-90-3, unii-v561v90bg2, ueberchlorsaeure, perchlorsaeure, perchloric acid, hclo4, hyperchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchloric acid, 1L, CHEBI:29221, Colorless, VLTRZXGMWDSKGL-UHFFFAOYSA-N, 11325, 100.454