Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FKCJ435017
also known as FKCJ/4350/17
Price (MYR)
133.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
133.00
EA
Add to cart
Specifications
pH2
ColorColorless
CAS7647-01-0
Boiling Point100°C
PackagingHDPE plastic bottle
View more
ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol