Check Order Status
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
HDPE plastic bottle
Catalog number
FKCJ433017
also known as FKCJ/4330/17
Price (MYR)
200.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
200.00
EA
Add to cart
Specifications
Color
Colorless
CAS
7647-01-0
Boiling Point
100°C
Packaging
HDPE plastic bottle
Quantity
2.5 L
View more
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis