Check Order Status
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
10 L
2.5 L
5 L
Packaging:
View all
Bag in box
Catalog number
FKCJ432024
also known as FKCJ/4320/24
Price (MYR)
570.00
BOT
Add to cart
Quantity:
10 L
Packaging:
Bag in box
Request bulk or custom format
Price (MYR)
570.00
BOT
Add to cart
Specifications
Color
Colorless
CAS
7647-01-0
Boiling Point
100°C
Packaging
Bag in box
Quantity
10 L
View more
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis