Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Actual product may vary
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, acide chlorhydrique, hydrogen chloride hcl, hydrogen chloride, chlorohydric acid, chlorwasserstoff, hydrochloric acid, spirits of salt, muriatic acid, 1L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FKCJ432015
also known as FKCJ/4320/15
1 LHDPE plastic bottle
Catalog number FKCJ432015
also known as FKCJ/4320/15
Price (MYR)
116.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Price (MYR)
116.00
EA
Add to cart
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, acide chlorhydrique, hydrogen chloride hcl, hydrogen chloride, chlorohydric acid, chlorwasserstoff, hydrochloric acid, spirits of salt, muriatic acid, 1L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
ColorColorless
CAS7647-01-0
Boiling Point100°C
PackagingHDPE plastic bottle
Quantity1 L
Molecular FormulaClH
Physical FormLiquid
MDL NumberMFCD00011324
Synonymhydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
IUPAC Namechlorane
Molecular Weight (g/mol)36.46
PubChem CID313
ChEBICHEBI:17883
Formula Weight36.46g/mol
Unit SizeEA