Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
5 L
Packaging:
View all
HDPE plastic bottle
Catalog number FKCJ288015
also known as FKCJ/2880/15
Price (MYR)
128.00
BOT
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
128.00
BOT
Add to cart
Specifications
ColorColorless
pH10
Physical FormLiquid
Quantity1 L
MDL Number217672
View more
Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement
Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
  • Standard and color-coded buffer solutions
  • Manufactured to a tolerance of ±0.02 pH units
  • Accurately standardized and labelled with lot-specific pH at 20°C
  • All buffers are traceable to NIST and BS 1647 standard
  • Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
  • Standard buffers are available over the range pH2 to pH10
  • Color-coded buffer solutions are available over the range pH4, pH7, and pH