Check Order Status
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Element Search
Structure Search
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid
Have Questions?
Change view
Quantity:
1 L
500 mL
2.5 L
Catalog number
FKCJ284015
also known as FKCJ/2840/15
Price (MYR)
304.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
304.00
EA
Add to cart
Chemical Identifiers
CAS
7778-77-0
Specifications
CAS
7778-77-0
Color
Colorless
pH
6
Physical Form
Liquid
Quantity
1 L
View more
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid