Check Order Status
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Element Search
Structure Search
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
Catalog number
FKCJ283015
also known as FKCJ/2830/15
Price (MYR)
245.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
245.00
EA
Add to cart
Chemical Identifiers
CAS
877-24-7
Specifications
CAS
877-24-7
Color
Colorless
pH
5
Physical Form
Liquid
Quantity
1 L
View more
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement