Check Order Status
Acetic Acid, 1M (1N ≈6%), Pure, Ready To Use
Element Search
Structure Search
Acetic Acid, 1M (1N ≈6%), Pure, Ready To Use
Have Questions?
Catalog Number
Quantity
Packaging
FKCJ055017
also known as FKCJ/0550/17
2.5 L
HDPE plastic bottle
Catalog number
FKCJ055017
also known as FKCJ/0550/17
Price (MYR)
355.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
355.00
EA
Add to cart
Specifications
Color
Colorless
MDL Number
36152
Quantity
2.5 L
Packaging
HDPE plastic bottle
Physical Form
Liquid
View more