Check Order Status
Iodine, 99.5+%, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Iodine, 99.5+%, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
100 g
250 g
Catalog number
FKCI045053
also known as FKCI/0450/53
Price (MYR)
677.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
677.00
EA
Add to cart
Specifications
Boiling Point
185°C
Melting Point
113°C
CAS
7553-56-2
Color
Gray
pH
5.1
View more