Check Order Status
Hydrofluoric Acid, 40%, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Hydrofluoric Acid, 40%, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCH140017
also known as FKCH/1400/17
Price (MYR)
682.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
682.00
EA
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7664-39-3
Melting Point
-35°C
Boiling Point
105°C
View more
Hydrofluoric Acid, 40%, Certified AR for Analysis