Check Order Status
Hydrobromic Acid, 48%, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Hydrobromic Acid, 48%, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FKCH0950PB17
also known as FKCH/0950/PB17
Price (MYR)
426.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
426.00
BOT
Add to cart
Specifications
pH
lt;1
Color
Yellow
CAS
10035-10-6
Melting Point
-11°C
Boiling Point
126°C
View more