Check Order Status
Hydrobromic Acid, 48%, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Hydrobromic Acid, 48%, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
2.5 L
Catalog number
FKCH0900PB08
also known as FKCH/0900/PB08
Price (MYR)
151.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
151.00
EA
Add to cart
Specifications
pH
lt;1
Color
Yellow
CAS
10035-10-6
Melting Point
-11°C
Boiling Point
126°C
View more