Hexanes, ≈95% N-Hexane, for HPLC
Hexanes, ≈95% N-Hexane, for HPLC
Hexanes, ≈95% N-Hexane, for HPLC
Hexanes, ≈95% N-Hexane, for HPLC
Hexanes, ≈95% N-Hexane, for HPLC
C6H14, CAS Number-110-54-3, hexanen, hexan, heksan, esani, dipropyl, skellysolve b, hexyl hydride, n-hexane, gettysolve-b, hexanes, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
Packaging:
View all
Aluminum Can
Catalog number FKCH040621
also known as FKCH/0406/21
Price (MYR)
756.00
EA
Add to cart
Quantity:
5 L
Packaging:
Aluminum Can
Request bulk or custom format
Price (MYR)
756.00
EA
Add to cart
Specifications
Melting Point-95°C
CAS110-54-3
ColorColorless
Boiling Point69°C
Quantity5 L
View more
C6H14, CAS Number-110-54-3, hexanen, hexan, heksan, esani, dipropyl, skellysolve b, hexyl hydride, n-hexane, gettysolve-b, hexanes, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97