Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Amber Glass Bottle
Catalog number FKCH035517
also known as FKCH/0355/17
Price (MYR)
224.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
224.00
BOT
Add to cart
Specifications
Melting Point-95°C
CAS92112-69-1
ColorColorless
Boiling Point69°C
Quantity2.5 L
View more
C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C