Check Order Status
Hexamine, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Hexamine, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
Plastic powder jar
Catalog number
FKCH032053
also known as FKCH/0320/53
Price (MYR)
278.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
278.00
EA
Add to cart
Specifications
CAS
100-97-0
Color
White
pH
8.5 to 9.5
Packaging
Plastic powder jar
Molecular Formula
C6H12N4
View more
Hexamine, Certified AR for Analysis