Check Order Status
Heptane, ≈99% N-Heptane, for HPLC
Element Search
Structure Search
Heptane, ≈99% N-Heptane, for HPLC
C7H16, CAS Number-142-82-5, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, heptane, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900, CCCCCCC, 3.5, 48mbar at 20 deg.C, 0.4 mPaS at 20 deg.C
Have Questions?
Change view
Quantity:
1 L
Packaging:
Amber glass bottle
Catalog number
FKCH010615
also known as FKCH/0106/15
Price (MYR)
258.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
258.00
EA
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
142-82-5
Vapor Pressure
48mbar at 20°C
Viscosity
0.4 mPaS at 20°C
Vapor Density
3.5
View more
C7H16, CAS Number-142-82-5, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, heptane, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900, CCCCCCC, 3.5, 48mbar at 20 deg.C, 0.4 mPaS at 20 deg.C