Check Order Status
Formic Acid, 90%, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Formic Acid, 90%, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FKCF1820PB17
also known as FKCF/1820/PB17
Price (MYR)
545.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
545.00
BOT
Add to cart
Specifications
pH
2.1
CAS
64-18-6
Melting Point
8°C
Boiling Point
101°C
Color
Colorless
View more