Check Order Status
Formamide, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Formamide, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
Plastic-coated Glass Bottle
Catalog number
FKCF1551PB17
also known as FKCF/1551/PB17
Price (MYR)
580.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
580.00
EA
Add to cart
Specifications
pH
4 to 5
Melting Point
2°C
CAS
75-12-7
Boiling Point
210°C
Quantity
2.5 L
View more
Formamide, Certified AR for Analysis