Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™
CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
25 L
Packaging:
View all
HDPE plastic bottle
Catalog number FKCF1501PB17
also known as FKCF/1501/PB17
Price (MYR)
185.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
185.00
EA
Add to cart
Specifications
PackagingHDPE plastic bottle
CAS50-00-0
Vapor Pressure2mbar at 20°C
Vapor Density> 1.0
Physical FormLiquid
View more
CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C