Check Order Status
Tisab Solution, Pure, Fisher Chemical™
Actual product may vary
Tisab Solution, Pure, Fisher Chemical™
Tisab Solution, Pure, Fisher Chemical, Quantity: 5L, Packaging: Plastic container, Colorless, pH: 5 to 7, MDL Number: 148650, Physical Form: Liquid
Have Questions?
Catalog Number
Quantity
FKCF132521
also known as FKCF/1325/21
5 L
Catalog number
FKCF132521
also known as FKCF/1325/21
Price (MYR)
411.00
EA
Add to cart
Quantity:
5 L
Price (MYR)
411.00
EA
Add to cart
Tisab Solution, Pure, Fisher Chemical, Quantity: 5L, Packaging: Plastic container, Colorless, pH: 5 to 7, MDL Number: 148650, Physical Form: Liquid
Specifications
Color
Colorless
pH
5 to 7
Physical Form
Liquid
Quantity
5 L
Formulation
Sodium Chloride, Water, 1,2-Cyclohexanediamine, Sodium Acetate
MDL Number
148650
Packaging
Plastic container
Solubility Information
Miscible